Assay Details
| Gene Symbol: LOC102140322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584302.2 | XP_005584359.1 | 2 - 3 | 2050 | 117 | |
| XM_005584303.2 | XP_005584360.1 | 2 - 3 | 2050 | 117 | ||
| XM_005584304.2 | XP_005584361.1 | 3 - 4 | 939 | 117 | ||
| XM_005584305.2 | XP_005584362.1 | 2 - 3 | 1256 | 117 | ||
More Information
Set Membership: |
|

LOC102140322
Genomic Map