Assay Details
Gene Symbol: INKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542349.2 | XP_005542406.1 | 1 - 2 | 231 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005542349.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
