Assay Details
| Gene Symbol: PCDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558077.2 | XP_005558134.1 | 3 | 4327 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDH1| Gene Symbol: PCDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558077.2 | XP_005558134.1 | 3 | 4327 | 98 | |
Set Membership: |
|