Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549538.2 | XP_005549595.1 | 3 | 593 | 170 | |
| XM_005549539.2 | XP_005549596.1 | 3 | 852 | 170 | ||
| XM_005549540.2 | XP_005549597.1 | 3 | 594 | 170 | ||
More Information
Set Membership: |
|

MRPS17
Genomic Map