Assay Details
| Gene Symbol: CEACAM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589366.2 | XP_005589423.1 | 2 | 1786 | 105 | |
| XM_005589367.2 | XP_005589424.1 | 2 | 1786 | 105 | ||
| XM_015442084.1 | XP_015297570.1 | 2 | 1788 | 105 | ||
More Information
Set Membership: |
|

CEACAM8
Genomic Map