Assay Details
| Gene Symbol: HINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557688.2 | XP_005557745.1 | 1 | 209 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005557688.2 transcripts.
Set Membership: |
|

HINT1
Genomic Map