Assay Details
| Gene Symbol: TOMM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539750.2 | XP_005539807.1 | 5 | 3346 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOMM20| Gene Symbol: TOMM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539750.2 | XP_005539807.1 | 5 | 3346 | 135 | |
Set Membership: |
|