Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592313.2 | XP_005592370.1 | 2 - 3 | 336 | 83 | |
| XM_005592314.2 | XP_005592371.1 | 2 - 3 | 278 | 83 | ||
More Information
Set Membership: |
|

LCAT
Genomic Map