Assay Details
| Gene Symbol: LOC102115795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579483.2 | XP_005579540.1 | 4 - 5 | 658 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102115795| Gene Symbol: LOC102115795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579483.2 | XP_005579540.1 | 4 - 5 | 658 | 78 | |
Set Membership: |
|