Assay Details
| Gene Symbol: LOC102121561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562701.2 | XP_005562758.1 | 2 | 649 | 107 | |
| XM_005562702.2 | XP_005562759.2 | 4 | 879 | 107 | ||
More Information
Set Membership: |
|

LOC102121561
Genomic Map