Assay Details
| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578900.2 | XP_005578957.1 | 1 - 2 | 743 | 95 | |
| XM_015435384.1 | XP_015290870.1 | 2 - 3 | 400 | 95 | ||
More Information
Set Membership: |
|

HBE1
Genomic Map