Assay Details
| Gene Symbol: MAFA-F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284601.1 | NP_001271530.1 | 2 - 3 | 270 | 104 | |
| XM_005553688.2 | XP_005553745.1 | 2 - 3 | 365 | 104 | ||
| XM_015448877.1 | XP_015304363.1 | 1 - 2 | 126 | 104 | ||
| GenBank mRNA | DQ367725.1 | - | 1 - 2 | 63 | 104 | |
| FS580595.1 | - | 1 - 2 | 93 | 104 | ||
More Information
Set Membership: |
|

MAFA-F
Genomic Map