Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563818.2 | XP_005563875.1 | 4 - 5 | 1033 | 106 | |
| XM_005563820.2 | XP_005563877.1 | 4 - 5 | 935 | 106 | ||
| XM_005563821.1 | XP_005563878.1 | 4 - 5 | 736 | 106 | ||
| XM_005563822.2 | XP_005563879.1 | 4 - 5 | 689 | 106 | ||
| XM_015455084.1 | XP_015310570.1 | 6 - 7 | 1112 | 106 | ||
| XM_015455085.1 | XP_015310571.1 | 5 - 6 | 1003 | 106 | ||
| XM_015455086.1 | XP_015310572.1 | 5 - 6 | 913 | 106 | ||
More Information
Set Membership: |
|

YWHAZ
Genomic Map