Assay Details
| Gene Symbol: RAB33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555944.2 | XP_005556001.1 | 1 - 2 | 650 | 82 | |
| XM_015450849.1 | XP_015306335.1 | 2 - 3 | 385 | 82 | ||
More Information
Set Membership: |
|

RAB33B
Genomic Map