Assay Details
| Gene Symbol: NLRP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434196.1 | XP_015289682.1 | 3 - 4 | 1151 | 96 | |
| XM_015434197.1 | XP_015289683.1 | 3 - 4 | 1148 | 96 | ||
| XM_015434198.1 | XP_015289684.1 | 3 - 4 | 1155 | 96 | ||
More Information
Set Membership: |
|

NLRP6
Genomic Map