Assay Details
| Gene Symbol: NLRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015429957.1 | XP_015285443.1 | 10 - 11 | 3480 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP3| Gene Symbol: NLRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015429957.1 | XP_015285443.1 | 10 - 11 | 3480 | 111 | |
Set Membership: |
|