Assay Details
| Gene Symbol: CASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284835.1 | NP_001271764.1 | 6 - 7 | 685 | 74 | |
| GenBank mRNA | AB220485.1 | - | 6 - 7 | 685 | 74 | |
| AY650318.1 | - | 5 - 6 | 580 | 74 | ||
More Information
Set Membership: |
|

CASP3
Genomic Map