Assay Details
| Gene Symbol: LOC102143631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441708.1 | XP_015297194.1 | 7 - 8 | 1663 | 96 | |
| XM_015441709.1 | XP_015297195.1 | 7 - 8 | 1660 | 96 | ||
More Information
Set Membership: |
|

LOC102143631
Genomic Map