Assay Details
| Gene Symbol: TAS2R13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570143.1 | XP_005570200.1 | 1 | 1202 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R13| Gene Symbol: TAS2R13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570143.1 | XP_005570200.1 | 1 | 1202 | 80 | |
Set Membership: |
|