Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454046.1 | XP_015309532.1 | 3 - 4 | 943 | 82 | |
| XM_015454047.1 | XP_015309533.1 | 3 - 4 | 442 | 82 | ||
More Information
Set Membership: |
|

DIO2
Genomic Map