Assay Details
| Gene Symbol: MARCHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556218.2 | XP_005556275.2 | 7 | 7655 | 75 | |
| XM_005556220.2 | XP_005556277.1 | 8 | 4804 | 75 | ||
| XM_005556221.2 | XP_005556278.1 | 8 | 5513 | 75 | ||
| XM_015450993.1 | XP_015306479.1 | 9 | 6248 | 75 | ||
| XM_015450994.1 | XP_015306480.1 | 8 | 5454 | 75 | ||
More Information
Set Membership: |
|

MARCHF1
Genomic Map