Assay Details
| Gene Symbol: ALPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586573.2 | XP_005586630.1 | 4 | 1356 | 93 | |
| XM_015439718.1 | XP_015295204.1 | 4 | 1355 | 93 | ||
| XM_015439719.1 | XP_015295205.1 | 3 | 887 | 93 | ||
More Information
Set Membership: |
|

ALPK2
Genomic Map