Assay Details
| Gene Symbol: CSGALNACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565039.2 | XP_005565096.1 | 6 - 7 | 1637 | 80 | |
| XM_015455958.1 | XP_015311444.1 | 5 - 6 | 2424 | 80 | ||
More Information
Set Membership: |
|

CSGALNACT2
Genomic Map