Assay Details
| Gene Symbol: IL15RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455518.1 | XP_015311004.1 | 378 | 97 | ||
| GenBank mRNA | FJ222748.1 | - | 205 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL15RA| Gene Symbol: IL15RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455518.1 | XP_015311004.1 | 378 | 97 | ||
| GenBank mRNA | FJ222748.1 | - | 205 | 97 | ||
Set Membership: |
|