Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284046.1 | NP_001270975.1 | 10 - 11 | 1099 | 82 | |
| XM_005573433.2 | XP_005573490.1 | 9 - 10 | 1032 | 82 | ||
| GenBank mRNA | AB170808.1 | - | 10 - 11 | 1099 | 82 | |
| AB170827.1 | - | 10 - 11 | 1094 | 82 | ||
More Information
Set Membership: |
|

SSB
Genomic Map