Assay Details
| Gene Symbol: CHORDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283229.1 | NP_001270158.1 | 7 - 8 | 645 | 110 | |
| XM_005579341.2 | XP_005579398.1 | 7 - 8 | 695 | 110 | ||
| XM_015435338.1 | XP_015290824.1 | 8 - 9 | 637 | 110 | ||
| XM_015435339.1 | XP_015290825.1 | 7 - 8 | 587 | 110 | ||
| GenBank mRNA | AB168720.1 | - | 7 - 8 | 645 | 110 | |
More Information
Set Membership: |
|

CHORDC1
Genomic Map