Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544660.2 | XP_005544717.1 | 3 - 4 | 582 | 72 | |
| XM_005544661.2 | XP_005544718.1 | 3 - 4 | 579 | 72 | ||
| XM_005544662.2 | XP_005544719.1 | 2 - 3 | 532 | 72 | ||
| XM_005544663.2 | XP_005544720.1 | 2 - 3 | 529 | 72 | ||
More Information
Set Membership: |
|

SZRD1
Genomic Map