Assay Details
| Gene Symbol: GJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283762.1 | NP_001270691.1 | 2 | 876 | 92 | |
| GenBank mRNA | AB169817.1 | - | 2 | 876 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GJA1| Gene Symbol: GJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283762.1 | NP_001270691.1 | 2 | 876 | 92 | |
| GenBank mRNA | AB169817.1 | - | 2 | 876 | 92 | |
Set Membership: |
|