Assay Details
| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566730.2 | XP_005566787.1 | 2 - 3 | 1752 | 92 | |
| XM_015456856.1 | XP_015312342.1 | 1 - 2 | 2237 | 92 | ||
More Information
Set Membership: |
|

INSYN2A
Genomic Map