Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558487.2 | XP_005558544.1 | 2 - 3 | 2350 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSYN2B| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558487.2 | XP_005558544.1 | 2 - 3 | 2350 | 102 | |
Set Membership: |
|