Assay Details
| Gene Symbol: EDDM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560711.2 | XP_005560768.1 | 1 - 2 | 124 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005560711.2 transcripts.
Set Membership: |
|

EDDM3B
Genomic Map