Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439226.1 | XP_015294712.1 | 3 - 4 | 2319 | 111 | |
| XM_015439227.1 | XP_015294713.1 | 4 - 5 | 2347 | 111 | ||
| XM_015439228.1 | XP_015294714.1 | 5 - 6 | 901 | 111 | ||
| XM_015439229.1 | XP_015294715.1 | 5 - 6 | 489 | 111 | ||
| XM_015439230.1 | XP_015294716.1 | 4 - 5 | 2330 | 111 | ||
| XM_015439231.1 | XP_015294717.1 | 4 - 5 | 2347 | 111 | ||
| XM_015439232.1 | XP_015294718.1 | 4 - 5 | 2347 | 111 | ||
More Information
Set Membership: |
|

CCDC122
Genomic Map