Assay Details
| Gene Symbol: COMMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564421.2 | XP_005564478.1 | 1 - 2 | 103 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005564421.2 transcripts.
Set Membership: |
|

COMMD5
Genomic Map