Assay Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319405.1 | NP_001306334.1 | 7 - 8 | 1126 | 59 | |
| GenBank mRNA | AB168793.1 | - | 7 - 8 | 1128 | 59 | |
More Information
Set Membership: |
|

DPEP3
Genomic Map