Assay Details
| Gene Symbol: FUT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552326.2 | XP_005552383.1 | 1 - 2 | 241 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005552326.2 transcripts.
Set Membership: |
|

FUT9
Genomic Map