Assay Details
| Gene Symbol: FUT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552326.2 | XP_005552383.1 | 2 - 3 | 325 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005552326.2 transcripts.
Set Membership: |
|

FUT9
Genomic Map