Assay Details
| Gene Symbol: LRRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548546.2 | XP_005548603.1 | 2 | 965 | 87 | |
| XM_005548547.2 | XP_005548604.1 | 2 | 1426 | 87 | ||
| XM_005548548.2 | XP_005548605.1 | 2 | 965 | 87 | ||
More Information
Set Membership: |
|

LRRC3
Genomic Map