Assay Details
| Gene Symbol: WDR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588127.2 | XP_005588184.1 | 5 - 6 | 609 | 76 | |
| XM_015440315.1 | XP_015295801.1 | 5 - 6 | 609 | 76 | ||
More Information
Set Membership: |
|

WDR83
Genomic Map