Assay Details
| Gene Symbol: DDIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571332.2 | XP_005571389.1 | 3 - 4 | 322 | 92 | |
| XM_005571333.2 | XP_005571390.1 | 3 - 4 | 267 | 92 | ||
| XM_005571334.2 | XP_005571391.1 | 3 - 4 | 736 | 92 | ||
| XM_015431103.1 | XP_015286589.1 | 1 - 2 | 935 | 92 | ||
More Information
Set Membership: |
|

DDIT3
Genomic Map