Assay Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582256.2 | XP_005582313.1 | 6 - 7 | 1186 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPP| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582256.2 | XP_005582313.1 | 6 - 7 | 1186 | 90 | |
Set Membership: |
|