Assay Details
| Gene Symbol: C12H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573814.2 | XP_005573871.1 | 1 - 2 | 1305 | 70 | |
| XM_015432525.1 | XP_015288011.1 | 2 - 3 | 734 | 70 | ||
More Information
Set Membership: |
|

C12H2orf66
Genomic Map