Assay Details
| Gene Symbol: MTRNR2L7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565000.1 | XP_005565057.1 | 1 | 997 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTRNR2L7| Gene Symbol: MTRNR2L7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565000.1 | XP_005565057.1 | 1 | 997 | 107 | |
Set Membership: |
|