Assay Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554987.2 | XP_005555044.1 | 2 - 3 | 2324 | 72 | |
| XM_005554988.2 | XP_005555045.1 | 2 - 3 | 2324 | 72 | ||
More Information
Set Membership: |
|

CCNI
Genomic Map