Assay Details
Gene Symbol: C3H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548985.2 | XP_005549042.1 | 2 - 3 | 235 | 98 | |
GenBank mRNA | AB170407.1 | - | 3 - 4 | 395 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
