Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578500.2 | XP_005578557.1 | 1 - 2 | 1602 | 57 | |
| XM_015435005.1 | XP_015290491.1 | 1 - 2 | 1602 | 57 | ||
More Information
Set Membership: |
|

NCR3LG1
Genomic Map