Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543009.2 | XP_005543066.1 | 3 - 4 | 404 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FPGT| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543009.2 | XP_005543066.1 | 3 - 4 | 404 | 100 | |
Set Membership: |
|