Assay Details
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588355.2 | XP_005588412.1 | 2 - 3 | 587 | 76 | |
| XM_005588356.2 | XP_005588413.1 | 2 - 3 | 299 | 76 | ||
More Information
Set Membership: |
|

NR2F6
Genomic Map