Assay Details
| Gene Symbol: RFX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284015.1 | NP_001270944.1 | 4 - 5 | 361 | 81 | |
| XM_005541901.2 | XP_005541958.1 | 3 - 4 | 309 | 81 | ||
| XM_005541902.2 | XP_005541959.1 | 4 - 5 | 454 | 81 | ||
| XM_005541903.2 | XP_005541960.1 | 4 - 5 | 296 | 81 | ||
| XM_005541905.2 | XP_005541962.1 | 4 - 5 | 645 | 81 | ||
| XM_005541906.2 | XP_005541963.1 | 4 - 5 | 369 | 81 | ||
| XM_005541907.2 | XP_005541964.1 | 4 - 5 | 802 | 81 | ||
| XM_015451209.1 | XP_015306695.1 | 3 - 4 | 890 | 81 | ||
| XM_015451211.1 | XP_015306697.1 | 3 - 4 | 530 | 81 | ||
| XM_015451216.1 | XP_015306702.1 | 3 - 4 | 485 | 81 | ||
| XM_015451219.1 | XP_015306705.1 | 3 - 4 | 617 | 81 | ||
| XM_015451222.1 | XP_015306708.1 | 4 - 5 | 735 | 81 | ||
| XM_015451224.1 | XP_015306710.1 | 4 - 5 | 362 | 81 | ||
| XM_015451229.1 | XP_015306715.1 | 4 - 5 | 742 | 81 | ||
| XM_015451233.1 | XP_015306719.1 | 4 - 5 | 442 | 81 | ||
| GenBank mRNA | AB168311.1 | - | 4 - 5 | 362 | 81 | |
More Information
Set Membership: |
|

RFX5
Genomic Map