Assay Details
| Gene Symbol: ZNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583398.2 | XP_005583455.1 | 7 - 8 | 1171 | 89 | |
| XM_005583399.2 | XP_005583456.1 | 7 - 8 | 1171 | 89 | ||
| XM_005583400.2 | XP_005583457.1 | 7 - 8 | 1171 | 89 | ||
| XM_005583401.2 | XP_005583458.1 | 6 - 7 | 1123 | 89 | ||
| XM_005583402.2 | XP_005583459.1 | 7 - 8 | 1171 | 89 | ||
| XM_015438151.1 | XP_015293637.1 | 7 - 8 | 1171 | 89 | ||
| XM_015438152.1 | XP_015293638.1 | 7 - 8 | 1171 | 89 | ||
| XM_015438153.1 | XP_015293639.1 | 6 - 7 | 1123 | 89 | ||
| XM_015438154.1 | XP_015293640.1 | 7 - 8 | 1492 | 89 | ||
More Information
Set Membership: |
|

ZNF207
Genomic Map