Assay Details
| Gene Symbol: NRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553629.2 | XP_005553686.1 | 1 - 2 | 1214 | 69 | |
| XM_015449671.1 | XP_015305157.1 | 1 - 2 | 1218 | 69 | ||
| XM_015449673.1 | XP_015305159.1 | 1 - 2 | 172 | 69 | ||
| XM_015449674.1 | XP_015305160.1 | 1 - 2 | 1220 | 69 | ||
More Information
Set Membership: |
|

NRM
Genomic Map